Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Osmometers

Osmometers determine the total solute particle concentration in physiological and biological liquids. Using the common freezing point depression method, the osmolality is reached by introducing a serum sample to the cooling chamber to reach a temperature below the expected freezing point. Then the sample is stirred for partial crystallization. The on-board computers in osmometers have the capability to recall a set number of past tests for easy comparisons.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"3"
  List View Easy View
Sort by:
 
 
 
 

Catalog Number: (76193-112)

Supplier:  Advanced Instruments
Description:   Best in class single-sample micro-osmometer that is easy-to-use offers precise, accurate results.
Supplier:  Advanced Instruments
Description:   The Model 210 Micro Osmometer determines the osmolality of solutions using freezing point depression (FPD).

Supplier:  Advanced Instruments
Description:   This single-sample osmometer utilizes the freezing-point depression method with 0.25mL samples, making it ideal for many clinical and industrial appli...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 3  of 3
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping