Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

ELISAs

https://ca.vwr.com/stibo/smallweb/std.lang.all/51/65/18945165.jpg

Enzyme-linked immunosorbent assays identify and calculate substances such as proteins, peptides, antibodies, and hormones. The provided reagents and supplies allow personnel to combine an immobilized antigen with an antibody that is joined to an enzyme. Following with substrate incubation, the produced antigen-antibody interaction and following conjugated enzyme activity can be measured and assessed. The ELISA kits are available for classroom simulations as well as for professional laboratory purposes.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"979"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalog Number: (CA95019-114L)
Supplier:  GE Healthcare - Life Sciences
Description:   ELISA.Specific for: free pro-MMP-9 and pro-MMP-9 complexed to TIMP-1. No cross-reactivity with pro-MMP-1, -2, -3, TIMP-1 and -2Range: 132 ng/ml.Sensitivity: 0.6 ng/ml.Suitable for use with cell culture supernatant and plasma samples.~ 4-h protocol.96 wells
Supplier:  Enzo Life Sciences
Description:   For the quantitative determination of testosterone in culture supernatants, plasma, serum, and saliva from any species
Catalog Number: (CA421101-BL)
Supplier:  Biolegend
Description:   The TMB Substrate Reagent Set contains 100 mL both Substrate Reagent A and Substrate Reagent B, sufficient for twenty 96-well plates
Supplier:  Enzo Life Sciences
Description:   Highly sensititive ELISA, meausring 8-iso-Prostaglandin F2α in plasma, serum and tissue.
Catalog Number: (75786-320)
Supplier:  Enzo Life Sciences
Description:   Quantitative determination of human, dog, pig, rabbit, bovine and horse-soluble LBP levels.
SDS
Supplier:  Enzo Life Sciences
Description:   The Corticosterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours
Supplier:  Promocell
Description:   The ox-LDL Enzyme-Linked-Immuno-Sorbent-Assay (ELISA) Kit is intended for the quantitative determination of ox-LDL in EDTA plasma, Heparin-plasma and ...
Supplier:  Biolegend
Description:   IL-3 ELISA MAX™ Deluxe ELISA Kit reacts with Rat
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 979
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next