Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Enzyme Assays

Either measuring substrate consumption or overall production, the enzyme assays are used to monitor enzymatic activity. Provided with the required supplies and reagents for any preferred method, enzyme kinetics and inhibition studies can be successfully completed. While continuous assay sampling methods give activity readings throughout development, discontinuous techniques calculate after the reaction has been stopped. The standardized enzyme assays will provide accurate identification and measurement of the protein molecules present in samples.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"233"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  LONZA PHARMA - BIOSCIENCE CA
Description:   The MycoAlert™ PLUS Mycoplasma Detection Kit is the next generation MycoAlert™ Kit. It provides higher light output to allow for use with less se...
SDS
Catalog Number: (CA89131-002)
Supplier:  G-Biosciences
Description:   G-Biosciences' β-Galactosidase Assay (CPRG) provides an easy, rapid, and highly sensitive method for determining the β-galactosidase activity in the l...
SDS
Supplier:  GE Healthcare - Life Sciences
Description:   Eliminate lengthy sample extraction procedures using direct assay technology
Supplier:  MOLECULAR DEVICES LLC. CA
Description:   IMAP® technology provides a homogeneous assay applicable to a wide variety of kinases without regard for substrate peptide sequence.
SDS
Supplier:  G-Biosciences
Description:   Methylation of key biological molecules and proteins plays important roles in numerous biological systems, including signal transduction, biosynthesis...
SDS
Supplier:  G-Biosciences
Description:   G-Biosciences' Fluorescent β-Galactosidase Assay (MUG) is a highly sensitive, fluorescent assay for determining the β-galactosidase activity in the ly...
SDS
Supplier:  Enzo Life Sciences
Description:   Biomol Green* Reagent 1 L^
Supplier:  Genlantis
Description:   For ultra-sensitive and quantitative measurement of low-level β-galactosidase expression.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 233
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next