Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • SBE (1) SBE (2)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "decontaminants"

https://ca.vwr.com/stibo/smallweb/std.lang.all/51/00/18945100.jpg

The decontaminants work instantly to trap and prevent the spread of micro-organisms. Commonly used in medical and research facilities, these cleaning agents remove, inactivate, or completely destroy contaminants. This makes items safe for handling and prevents diseases from spreading. While meeting safe working environment standards, the use of decontaminants will not result in any residue that would affect procedures. Solutions are versatile to efficiently clean all surfaces and equipment pieces.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"10"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalog Number: (10798-804)
Supplier:  Foxwest
Description:   ELIMINase removes RNase, DNase, unwanted DNA and radioactive contamination.
SDS
Supplier:  Decon Labs
Description:   NoCount® aerosol spray removes radioactive contaminants from hard surfaces. NoCount's foaming action helps to lift and trap particles so they can be e...
SDS
Supplier:  Thermo Scientific
Description:   This simple, safe and economical disposal system for both 10% buffered and unbuffered formalin eliminates the need for expensive waste hauling and ons...
Supplier:  Thermo Scientific
Description:   RNase AWAY® eliminates RNase and DNA from laboratory surfaces. The surface decontaminant is ideal for decontaminating apparatus, benchtops, glassware,...
SDS
Supplier:  Biodex Medical Systems
Description:   For safely removing radioactive contaminants from skin, cloth, metal, glass, floors, plastic, and equipment. Will also remove general contaminants, su...
SDS
Small Business Enterprise
Supplier:  Thermo Scientific
Description:   RBS 35 Detergent Concentrate is a general cleaning agent for use on many laboratory surface materials and glassware, including lenses, mirrors, and re...
SDS
Supplier:  Foxwest
Description:   RNase Displace removes RNase and unwanted DNA. It works instantly, eliminating the time-consuming method of autoclaving. It can be used on all surface...
Supplier:  Argos Technologies
Description:   Surface decontaminant specially formulated to eliminate RNase and DNase from laboratory glassware, plastics, stainless steel, and other surfaces.
Small Business Enterprise
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 10
  1  2  Next