Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Healthcare Controls

https://ca.vwr.com/stibo/smallweb/std.lang.all/93/87/18989387.jpg

Screening equipment commonly used in the medical field, ready-to-use heathcare controls compare measurements to validate test results. Designed for specific condition analyses, the stable reagents and supplies come in different levels for accurate determinations. Bottled for easy dispensing and longtime storage, the versatile products are assayed for compatibility with automated instruments or traditional methods. By standardizing examinations, heathcare controls guarantee accurate diagnoses are reported to patients for the correct treatment commencement.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"21"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Thermo Scientific
Description:   Identify random or systematic errors in nucleic acid test procedures for hepatitis C viral RNA (HCV RNA) with these controls.
Limited Availability
Supplier:  Thermo Scientific
Description:   Reagents must be stored at 2 to 8°C (36 to 46°F)
SDS
Supplier:  Azer Scientific Inc.
Description:   Linearity controls are liquid stable, ready-to-use reagents for use with all major brands of blood gas, electrolyte, and chemistry analyzers.
Product available on GSA Advantage® Minority or Woman-Owned Business Enterprise
Supplier:  Quantimetrix
Description:   THE DROPPER PLS URINE DIP BX10
SDS
Supplier:  Wako Life Sciences
Description:   The Autokit CH50 is an in vitro liposome immunoassay (LIA) for the quantitative determination of total complement activity (CH50) in human serum, usin...
Catalog Number: (CA97000-018)
Supplier:  Wako Life Sciences
Description:   The Cholesterol E is an enymatic colorimetric method for the quantitative determination of total cholesterol in serum.
Supplier:  Quantimetrix
Description:   SWEAT CTRL SET,LVL1,2,3,9X3ML
SDS
Supplier:  YSI
Description:   Zobell solution is used for calibrating ORP/Redox on several different instruments.
SDS
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 21
  1  2  3  Next