Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Benchtop Coolers

https://ca.vwr.com/stibo/smallweb/std.lang.all/49/37/18944937.jpg

Compact enough to fit any work countertop, the benchtop coolers preserve the integrity of biological samples. Maintaining freezing conditions longer, the insulated containers safely stop reactions, store, and transport temperature sensitive materials. With quality engineering, these cooling systems provide a more convenient method to minimize enzyme, cell, reagent, or solution loss. Benchtop coolers hold standard tube and vial sizes in various matrixes and may feature labeled grids for fast identification.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"37"
  List View Easy View
Sort by:
 
 
 
 

Catalog Number: (414004-284)

Supplier:  VWR International
Description:   When filled with isopropyl alcohol and placed in mechanical freezer, this polycarbonate freeze controller provides the -1°C per minute cooling rate re...
Supplier:  VWR International
Description:   Coolers hold 0.5 or 1.5 mL microcentrifuge tubes at or below -20°C (-4°F) for up to one hour on the benchtop or in transport.
Supplier:  VWR International
Description:   These reversible benchtop chillers hold either 0.5 or 0.65 mL tubes on one side and 1.5 to 2.0 mL tubes on the other.
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product is manufactured with low CO₂ impact
Supplier:  VWR International
Description:   Racks float in your water bath and can accommodate dry ice for use with frozen samples on the bench.
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product is manufactured with low CO₂ impact
Catalog Number: (89004-570)

Supplier:  VWR International
Description:   Benchtop rack allows frozen PCR samples to be worked with for up to four hours at 0°C (32°F).
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product is manufactured with low CO₂ impact
Supplier:  VWR International
Description:   The VWR® Benchtop Cooler is an excellent alternative to messy ice buckets and water baths which can be a source of contamination.
Product available on GSA Advantage®
Supplier:  Heathrow Scientific
Description:   Protect temperature-sensitive samples at the bench or in the freezer.
Supplier:  Heathrow Scientific
Description:   Durable containers for chilling temperature-sensitive samples.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 37
  1  2  3  4  5  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping