Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

chest-coolers

https://ca.vwr.com/stibo/smallweb/std.lang.all/50/20/18945020.jpg

The leak-proof chest coolers are insulated for the best temperature retention when contents are sensitive to environmental changes. Hinged or detachable lids prevent cooling agents from escaping to extend the duration that containers can provide temperature stability. Handles may come in molded, towing, overhead, or dual styles based on user preference. Chest coolers are available in bright colors with or without the additional wheel and channel drain features to improve transportation and emptying.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"8"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Therapak Corporation
Description:   Coolers feature generous foam insulation, along with hinged lids and lid-locking handles for safe transport of contents.
Supplier:  Therapak Corporation
Description:   Coolers feature detachable lids and a wide bottom base for extra stability.
Supplier:  Therapak Corporation
Description:   Coolers feature two-way handles for easy lifting, hinged lids, and rustproof, leak-resistant, no-tilt drains. The insulation is environmentally friend...
Supplier:  Therapak Corporation
Description:   Coolers feature environmentally friendly insulation with no ozone-depleting materials. They have molded handles for easy carrying and removable lids.
Supplier:  Therapak Corporation
Description:   Coolers have extra insulation to provide superior cold retention, and can hold ice for up to five days at temperatures up to 90°F. They are equipped w...
Catalog Number: (89171-062)
Supplier:  Therapak Corporation
Description:   Rustproof, leak-resistant coolers feature large tow handles and wheels. They are equipped with no-tilt drains and environmentally friendly insulation....
Catalog Number: (89171-100)
Supplier:  Therapak Corporation
Description:   Rustproof, leak-resistant coolers are equipped with no-tilt drains and 5.08cm (2”) of insulation built into the lid. Coolers feature two-way handles f...
Catalog Number: (14228-934)
Supplier:  Techne
Description:   The cooling head of the Techne® Dip Cooler fits neatly and unobtrusively into the corner of the bath and can be secured with a specially designed...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 8