Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Fall Lanyards and Ropes

https://ca.vwr.com/stibo/smallweb/std.lang.all/52/17/18945217.jpg

Lanyards and ropes for use with fall harnesses or belts are available in fixed lengths, self-retracting styles, or stretchable, shock-absorbing lanyards. Expanding lanyards reduce the fall arrest force impact on the individual, allow for great maneuverability, and reduce the chances of tripping, snagging, or dragging. Lanyards are lightweight but possess great carrying capacity. The heavy-duty materials used in manufacturing these products provide excellent abrasion resistance to keep workers safely suspended regardless of encountered surroundings.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Honeywell Safety Products
Description:   Alleviates the effects of orthostatic intolerance, also known as suspension trauma.
Supplier:  Honeywell Safety Products
Description:   This all-in-one lanyard features SoftStop shock absorber and a cross-arm anchorage connector specially designed for tie-back use
Supplier:  Honeywell Safety Products
Description:   For securing to I-beams and other structures
Supplier:  Honeywell Safety Products
Description:   Lanyards feature super-strong outer jacket with shock-absorbing inner core to reduce fall arrest forces below 480kg (900lbs.)
CSA Certified
Supplier:  Honeywell Safety Products
Description:   Stretchable lanyards feature a specially woven, shock-absorbing inner core that expands up to 1.1m (3.6') to reduce fall arrest forces to 408kg (900lb...
CSA Certified
Catalog Number: (470089-222)
Supplier:  ROCKFORD MFG. CO.
Description:   ROPE CLOTHESLINE 7/32 X 50'
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 6  of 6
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping