Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Bell Jars

https://ca.vwr.com/stibo/smallweb/std.lang.all/95/12/18709512.jpg

Bell jars are used in conjunction with a vacuum fitted base to form and maintain a low-pressure environment. Ground flat rims provide the tightest seal for minimum leakage. Round bottom styles with the molded solid knob handles are easy to remove to access items. The glassware is optically clear for best experimenting visibility. Choose from various bell jar dimensions when requiring a low-quality vacuum atmosphere or simply protecting delicate materials from harmful surroundings.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"8"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalog Number: (470121-432)
Supplier:  VWR International
Description:   Lasts for Years!
Catalog Number: (14207-502)
Supplier:  Wilmad-LabGlass
Description:   This borosilicate glass bell jar has a plain dome and a rim that is ground flat for a tight seal.
Supplier:  Variety Glass
Description:   VWR BELL JAR W/GLASS STOPPER.
Supplier:  CORNING CA
Description:   These specially blown, sturdy, round-bottom jars have a fine ground finish at the open end
Product available on GSA Advantage®
Supplier:  CORNING CA
Description:   Tall form, glass bell jars with heavy, solid knob handle
Product available on GSA Advantage®
Supplier:  Variety Glass
Description:   This bell jar is made with heavy-duty flint glass.
Catalog Number: (470104-014)
Supplier:  Variety Glass
Description:   JAR BELL W/O STOPPER 13 CMX23 CM 2 L
Catalog Number: (470020-808)
Supplier:  AMERICAN EDUCATIONAL PROD
Description:   VACUUM PL-BELL JR W/FTG FOR .25 ID HOSE
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 8
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping