Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Magnifier Lamps

https://ca.vwr.com/stibo/smallweb/std.lang.all/94/96/18989496.jpg

Designed to allow precise work, magnifier lamps bring definition and contrast to objects under the viewport. Common uses for magnifier lamps include inspection, electronics, quality control, sample examination, and biological analysis. The large viewing areas make observations easy on the eyes and the highly polished glass of these lenses provides distortion-free viewing. Various sizes and clamps allow for desk mounting in larger workspaces or areas with limited amounts of space.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"4"
  List View Easy View
Sort by:
 
 
 
 

Catalog Number: (10048-662)

Supplier:  VWR International
Description:   Magnifier Lamp is the ideal tool to view objects at two times magnification in bright light, with high contrast and a sharply defined focus.
Supplier:  Kewaunee
Description:   Magnifier light features a 12.7cm (5"), 3-diopter double convex precision ground lens which produces distortion-free magnification under bright fluore...
Product available on GSA Advantage®
Catalog Number: (470004-888)

Supplier:  M.G. ELECTRONICS
Description:   Exceptional clarity at an affordable price.
Catalog Number: (82008-002)

Supplier:  Heathrow Scientific
Description:   The Mini Magnifier enlarges objects for easier viewing
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 4  of 4
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping