Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • GSA Contract (7) UL Listed (20)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Sale Items
  • Search Within Results

Measuring (Mohr) Pipets

https://ca.vwr.com/stibo/smallweb/std.lang.all/96/06/18709606.jpg

Measuring Mohr pipettes have uniform graduation intervals on the transparent stem only. With exact drop reproducibility from the tapered tip, the narrow tube devices support a controlled testing environment. Different drawing capacity options are activated by simply squeezing the bulb. Using the volumetric Mohr pipette, the final volume is calculated using the difference of liquid level before and after. Since the instruments can be difficult to sterilize, disposable options are available.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"29"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   VWR® pipettes are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
UL Listed
Supplier:  VWR International
Description:   VWR® Mohr pipets are made from Borosilicate glass, ASTM 438, Type 1, Class B standard
UL Listed
Supplier:  VWR International
Description:   VWR® pipettes are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
UL Listed
Supplier:  VWR International
Description:   RUBBER BULB PIPET 5ML 6/PK..
Supplier:  VWR
Description:   Designed for Consisent Drop Sizes
Catalog Number: (470104-202)

Supplier:  VWR
Description:   PIPET GRAD DROPPER FLNT GLS 178 MM 3 ML
Supplier:  VWR
Description:   These polyethylene pipets are chemically inert.
Catalog Number: (470152-908)

Supplier:  VWR
Description:   Chemically inert polyethylene pipets. Designed for consistent bulb pressure and tip circumference to ensure precisely measured drop reproducibility.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 29
  1  2  3  4  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping