Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "combination-tube-racks"

https://ca.vwr.com/stibo/smallweb/std.lang.all/95/31/18709531.jpg

Versatile combination tube racks are capable of holding multiple sizes or shapes for maximum loading flexibility. Interlocking models connect in multiple orientations and can be simply flipped over to have a different diameter size hole upright. Available in different color options, select combination tube racks also support different laboratory equipment such as bottles or plates. Options with a tiered formation store a variety of tube sizes in the most compact way possible.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"17"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   These heavy-duty, polypropylene racks link together to meet custom rack needs.
Catalog Number: (89096-724)
Supplier:  VWR International
Description:   Expandable, interlocking four-part rack system will accommodate 15 mL and 50 mL centrifuge tubes, microcentrifuge tubes, PCR tubes and plates, and cul...
Product and/or packaging contains recycled/renewable content
Product and/or packaging is recyclable or reduces waste
Product is manufactured with low CO₂ impact
Catalog Number: (89234-744)
Supplier:  VWR International
Description:   Quickly change tube sizes with this adaptable polypropylene rack.
Catalog Number: (82010-746)
Supplier:  VWR International
Description:   These polypropylene racks can slide together in multiple orientations to work with different-sized tubes at the same time.
Limited Availability
Supplier:  VWR International
Description:   Racks allow loading and transportation of several sizes of test tubes, culture tubes, centrifuge tubes, and microcentrifuge tubes.
Catalog Number: (89140-272)
Supplier:  VWR International
Description:   Dilute or take off concentrate using one rack with the use of the Duo Tube Rack.
Supplier:  VWR International
Description:   VWR® Reagent Rack systems provide excellent storage of laboratory equipment above the lab station.
Supplier:  Bel-Art Products
Description:   Perfect for benchtop use or freezer storage, this double-sided polypropylene plastic rack accommodates 0.2 ml thin-walled PCR tubes on one side while ...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 17
  1  2  3  Next