Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "microbiology-stains"

https://ca.vwr.com/stibo/smallweb/std.lang.all/53/27/18945327.jpg

Microbiology stains are used to detect and identify microorganisms that would otherwise be difficult to find in clinical specimens. Simple positively or negatively charged dyes create a fast discovery. The premixed solutions artificially color to help observe movement, structure, and characteristics of live or preserved specimens. Differentiating microorganisms based on properties, color combination procedures like the Gram stain or acid-fast techniques require microbiology stains.

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
 
 
SearchResultCount:"35"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   Gram Stain Sets include all the necessary solutions to fully differentiate Gram positive and Gram negative specimens.
SDS
Supplier:  VWR
Description:   A colorimetric screening system, based on the expression of the lac operon’s ?-galactosidase coding gene lacZ, is widely used in the selection of bact...
SDS
Supplier:  MilliporeSigma
Description:   Stains are manufactured with materials that are certified by the Biological Stain Commission.
SDS
Supplier:  MilliporeSigma
Description:   Löffler's methylene blue solution is generally used as bacteriological counter staining of, e.g. of gonococcae, lactic acid bacteria and for visualiza...
SDS
Supplier:  MilliporeSigma
Description:   Certified biological stain.
SDS
Supplier:  HARDY DIAGNOSTICS CA
Description:   Crystal violet stain is used as a primary stain in gram staining procedures. The decolorizer is intermediate strength, comprised of 50% alcohol and 50...
Catalog Number: (CA89428-916)
Supplier:  HARDY DIAGNOSTICS CA
Description:   India Ink, for Cryptococcus, dropper bottle, 15ml, by Hardy Diagnostics, prefer to ship ground.

For use in the identification of Cryptococc...
Supplier:  Tonbo Biosciences
Description:   Ghost Dye™ Red 780 is an amine reactive viability dye that can be used to discriminate viable from non-viable mammalian cells in flow cytometry applic...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 35
  1  2  3  4  5  Next