Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "Prepared Microbiology Media"

https://ca.vwr.com/stibo/smallweb/std.lang.all/96/75/18989675.jpg

Prepared microbiology media support accurate microorganism detection, identification, and cultivation. Saving time by eliminating the requirement to complete the groundwork, the media increase the overall working efficiency for any laboratory. Minimizing the possibility of formula variability, the standardized solutions ensure that data errors do not occur. The prepared microbiology media are plated or contained in tubes to guarantee that the correct nutrient mixtures are present for specific cell types.

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
 
 
SearchResultCount:"336"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR
Description:   LURIA AGAR W/AMP.&60 PLATES (6)200ML BTL
Supplier:  IPM SCIENTIFIC, INC.
Description:   AGAR (COMPLETE YED) PKG/10 PLATES .
Supplier:  IPM SCIENTIFIC, INC.
Description:   This media can be used to determine if a microorganism is capable of hydrolyzing starch as a source of energy for growth.
Supplier:  IPM SCIENTIFIC, INC.
Description:   For Fermentation Studies
Catalog Number: (470180-660)
Supplier:  VWR
Description:   Contains 12 nutrient agar deeps. Used to check for organism motility. 17 mL per tube.
Catalog Number: (470180-650)
Supplier:  VWR
Description:   TUBES TRIPPLE SUGAR IRON (TSI) PKG/12
Supplier:  IPM SCIENTIFIC, INC.
Description:   For Maintaining Haploid Yeast Cultures
Supplier:  VWR
Description:   LURIA AGAR&60 PLATES (6)200ML BTL
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 336
  1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  Next