VWR® Critical Environment Swab, Small Foam Head
Supplier: VWR International
SU-070VFRTD
76478-848BX 156.77 CAD
76478-848
VWR® Critical Environment Swab, Small Foam Head
Swabs Swabs for Clinical Use
Swab features 100 ppi open cell polyurethane foam with 100% virgin polypropylene handle.
  • Polyurethane foam head
  • Virgin polypropylene handle
  • Small size head
Swab head is thermally bonded to the handle without using any adhesive.