VWR® Critical Environment Swab, Small Pointed Compressed Foam Head
Supplier: VWR International
SU-070VFSTD
76478-852BX 136.16 CAD
76478-852
VWR® Critical Environment Swab, Small Pointed Compressed Foam Head
Swabs Swabs for Clinical Use
Swab features 100 ppi open cell polyurethane foam with 100% virgin polypropylene handle.
  • Polyurethane foam head
  • Virgin polypropylene handle
  • Small pointed foam head
Swab head is thermally bonded to the handle using no adhesive.