Denhardt's solution concentrate ≥99.0%, OmniPur®
Supplier: MilliporeSigma
3610-50ML
CA-EM3610EA 248.73 CAD
CA-EM3610
Denhardt's solution concentrate ≥99.0%, OmniPur®
Buffers Electrophoresis Buffers Blocking Buffers
50X. Solution of 1.0% Ficoll*, 1.0% (w/v) Polyvinyl Pyrrolidone, 1.0% (w/v) Bovine Serum Albumin.
DNase-, RNase-, and protease-free.