Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

KIMAX® Media/Storage Bottles, KG-35 Borosilicate Glass, Graduated, Kimble Chase

Supplier: DWK Life Sciences (KIMBLE®)
KIMAX®
61110T-250 61110P-500 61110P-125 61110R-500 61110R-125 61110R-1000 61110P-1000 61111T-1000 61111P-1000 61110T-500 61110T-125 61110P-250 61110R-250 61110T-1000 61111R-1000
Screw thread graduated bottles come assembled with autoclavable closures

Choose from closures that are black phenolic 14-B white rubber-lined, black phenolic PTFE-faced 14-B white rubber-lined, or black polypropylene welded to a PTFE/silicone liner. The PTFE/silicone liner eliminates the possibility of glue contamination to bottle contents. All bottles feature a large white marking spot




 

ORDER

Expand All / Collapse All
Capacity Dimensions Cap Type Graduation Interval Graduation Range GPI Finish Supplier No. VWR Catalog Number Unit Price Quantity
125 mL (4.2 oz.) 55 O.D. x 123 H, mm Black Phenolic PTFE-Faced 14-B White Rubber-Lined Closure 25 mL 50–100 mL 38-430 61110T-125 16171-010 Case of 48 Retrieving
125 mL (4.2 oz.) 55 O.D. x 123 H, mm Black Phenolic 14-B White Rubber-Lined Closure 25 mL 50–100 mL 33-430 61110R-125 16171-004 Case of 48 Retrieving
125 mL (4.2 oz.) 55 O.D. x 123 H, mm Black Polypropylene Closure Welded to PTFE/Silicone Liner 25 mL 50–100 mL 33-430 61110P-125 66014-004 Case of 48 Retrieving
1 L (33.8 oz.) 102 O.D. x 226 H, mm Black Phenolic PTFE-Faced 14-B White Rubber-Lined Closure 50 mL 100–900 mL 33-430 61111T-1000 16171-016 Case of 12 Retrieving
1 L (33.8 oz.) 102 O.D. x 225 H, mm Black Phenolic PTFE-Faced 14-B White Rubber-Lined Closure 50 mL 100–900 mL 38-430 61110T-1000 16171-006 Case of 12 Retrieving
1 L (33.8 oz.) 102 O.D. x 225 H, mm Black Phenolic 14-B White Rubber-Lined Closure 50 mL 100–900 mL 38-430 61110R-1000 16171-000 Case of 12 Retrieving
1 L (33.8 oz.) 102 O.D. x 226 H, mm Black Phenolic 14-B White Rubber-Lined Closure 50 mL 100–900 mL 33-430 61111R-1000 16171-014 Case of 12 Retrieving
1 L (33.8 oz.) 102 O.D. x 225 H, mm Black Polypropylene Closure Welded to PTFE/Silicone Liner 50 mL 100–900 mL 33-430 61110P-1000 66014-010 Case of 12 Retrieving
250 mL (8.5 oz.) 70 O.D. x 150 H, mm Black Phenolic PTFE-Faced 14-B White Rubber-Lined Closure 25 mL 50–200 mL 33-430 61110T-250 66014-000 Case of 48 Retrieving
250 mL (8.5 oz.) 70 O.D. x 150 H, mm Black Phenolic 14-B White Rubber-Lined Closure 25 mL 50–200 mL 33-430 61110R-250 66014-002 Case of 48 Retrieving
250 mL (8.5 oz.) 70 O.D. x 150 H, mm Black Polypropylene Closure Welded to PTFE/Silicone Liner 25 mL 50–200 mL 33-430 61110P-250 66014-006 Case of 48 Retrieving
500 mL (16.9 oz.) 88 O.D. x 192 H, mm Black Polypropylene Closure Welded to PTFE/Silicone Liner 50 mL 100–400 mL 33-430 61110P-500 66014-008 Case of 24 Retrieving
500 mL (16.9 oz.) 88 O.D. x 192 H, mm Black Phenolic PTFE-Faced 14-B White Rubber-Lined Closure 50 mL 100–400 mL 33-430 61110T-500 16171-008 Case of 24 Retrieving
500 mL (16.9 oz.) 88 O.D. x 192 H, mm Black Phenolic 14-B White Rubber-Lined Closure 50 mL 100–400 mL 33-430 61110R-500 16171-002 Case of 24 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty