Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl ether, ACS/HPLC Certified, contains 2% ethanol perservative, Honeywell Research Chemicals

Supplier: HONEYWELL BURDICK & JACKSON CA
Synonyms: 2,2'-Oxybisethane2,2'-OxydiethaneEtherEthoxyethane

AH106-4PC AH106-4
For HPLC, spectrophotometry, and applications requiring ACS reagent grade solvents. UV cutoff at 215nm.


Formula: (CH₃CH₂)₂O
MW: 74,12 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: -123 °C
Density: 0.7135 g/cm³ (20 °C)
Flash Pt: -40 °C
MDL Number: MFCD00011646
CAS Number: 60-29-7
UN: 1155
ADR: 3,I
Merck Index: 12,03852
https://ca.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg
Specification Test Results
Water by Karl Fischer Titration 0.03% Max.
GC Analysis (excluding ethanol) 99.9% Min.
Color (APHA) 10 Max.
Refractive Index (20°C) 1.3523 Min. to 1.3533 Max.
UV Absorbance @ 300 nm 0.005 AU Max.
UV Absorbance @ 400 nm 0.005 AU Max.
UV Absorbance @ 275 nm 0.010 AU Max.
UV Absorbance @ 250 nm 0.080 AU Max.
Titrable Acid 0.2 µeq/g Max.
Peroxide (as H2O2) 1 ppm Max.
UV Cutoff 215 nm
Residue 3 mg/L
Meets ACS Specifications


 

ORDER

Expand All / Collapse All
Size Packaging Supplier No. VWR Catalog Number Unit Price Quantity
4 L Glass Bottle AH106-4 CABJAH106-4 Case of 4 (4 l) Retrieving Restricted
Each(4l) Retrieving Restricted
restricted You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty