VWR® Digital Outlet Controller

Supplier: VWR, part of Avantor
VWR®
5090
61161-343EA 183.25 CAD
61161-343
VWR® Digital Outlet Controller
Timers Time Switches
Versatile and economical controller programs equipment on and off at any minute of any day.

  • Traceable®
  • High-impact ABS plastic case
  • Motor load capacity: 1 hp

A fail-safe memory permits programming 14 different switchings per day.

The 1.3 cm (¹/₂") LCD shows the time-of-day, the day of the week, and the event number of the program taking place. Manual override button permits equipment to be switched on/off independent of event settings. Program and clock times are maintained by a backup battery in the event of an electrical outage.

They may be allocated over four separate weekly programs for up to 196 switchings per week. Controller will turn equipment on and off in intervals as short as 1 minute or as long as 168 hours. The a.m./p.m. day programs allow repeating the same cycle every day of the week, weekdays only, weekends only, or any selected days.

Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.

Ordering information: Includes two 1.5 V batteries and operating instructions.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR