PYREX® Filtering Funnels, 60° Angle, Corning®
Supplier: Corning Life Sciences
PYREX®
6180-50 6140-65 6120-6 6140-75 6100-6 6120-5 6180-65 6160-75 6140-50 6140-100 6160-65 6180-100 6180-75 6100-5
30231-024CS 1114.25 CAD
30231-024 30230-500 89090-548 30230-503 89090-546 89090-544 30231-046 30232-061 30230-929 30230-984 30232-050 30231-080 30231-068 89091-640
PYREX® Filtering Funnels, 60° Angle, Corning®
Funnels Filtration Funnels Filter Funnels
Made from hand-blown blanks with approximate 60° angle, these funnels are for use in flasks or test tubes without a filter rack
These filtering funnels are made of Pyrex® borosilicate glass.
Funnels feature a beaded edge for strength. Fluted funnels feature pressed inside flutings to further decrease filtering time.