VWR®, Beakers, PTFE
Catalog #: 89026-030
Supplier: VWR International
Price
360,50 $
Log in for availability
About this item
Isostatically molded in pure, FDA approved PTFE, these beakers feature a pouring spout and have a machined base for good heat transfer.
- Chemically resistant and inert
- Autoclavable
Machined base provides good heat transfer in these PTFE beakers.
Customers Who Bought This Also Bought