PYREX® Bushing-Type Adapter with [ST] Joint, Corning®
Supplier: Corning Life Sciences
PYREX®
8825-3424
89091-544CS 1984.32 CAD
89091-544
PYREX® Bushing-Type Adapter with [ST] Joint, Corning®
Adapters Adapters, Conical Ground Joints
Adapter features a heavy wall for mechanical strength, and a heavy, uniform rim at the top provides a serviceable finger grip.
  • Heavy wall
  • Rim at top
  • Shorter than conventional types
Adapter is shorter than the conventional types and permits more compact assemblies. Useful for attaching [ST] thermometers.