PYREX® Bell Jars, Corning
Supplier: Corning
Certificates
About this item
These specially blown, sturdy, round-bottom jars have a fine ground finish at the open end. Flat, ground surface has a contact area equal to the wall thickness at the open end.
Caution: Do not use with heat, pressure or vacuum applications.
Customers Who Bought This Also Bought