Cryogenic Vials with Closures, Polypropylene, Corning®
Supplier: Corning
430656 430489 430487 431386 430661 430488 430490 430659 430662 430492 430491 430658 430663
66021-970CS 971.32 CAD
66021-970 66021-944 66021-940 89089-764 66021-976 66021-942 66021-946 66021-974 66021-978 66021-950 66021-948 66021-972 66021-980
Cryogenic Vials with Closures, Polypropylene, Corning®
Vials Cryogenic Vials
These cryogenic vials are manufactured from polypropylene to withstand temperatures to –196 °C (–320.8 °F)
  • Large marking spot for easy labeling of contents
  • DNase- and RNase-free, and non-pyrogenic
All self-standing vials have a special base design allowing them to be locked into the Corning® cryogenic vial rack, Polypropylene, 50-Position for single-handed manipulation.
Ordering information: All vials except the external thread vial with plug seal cap can be color-coded with cap inserts, which are sold separately.
Choose from round or freestanding bottoms. Avalable with a variety of closures. Cap insert accessory in assorted colors includes 100 each of white, blue, red, green, and yellow.
Caution: Do not use cryogenic vials for storage in the liquid phase of liquid nitrogen.