ContactUsLinkComponent
Bilsom™ THUNDER® Headband Earmuffs, Honeywell Safety
Bilsom™ THUNDER® Headband Earmuffs, Honeywell Safety
Catalog #: CA89005-762
Bilsom™ THUNDER® Headband Earmuffs, Honeywell Safety
Catalog #: CA89005-762
Discontinued
Specifications
  • NRR:
    30 dB
  • Description:
    Earmuffs Thunder T3
  • Cat. no.:
    CA89005-762
Specifications
About this item
Earmuffs use Air Flow Technology™ to provide better, more consistent attenuation, especially at lower frequencies
  • Sturdy ABS plastic headband minimizes pressure on the head and withstands rough treatment while maintaining its shape
  • Soft, comfortable contact surface of the headband breathes easier in warm and humid climates
  • Broad snap-in ear cushions allow plenty of room for the outer ear and are easily replaced
Dielectric construction is suitable for electrical and mining environments.
Customers Who Bought This Also Bought