Polystyrene Spreaders
Catalog #: 47732-760
Supplier: Avantor
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
SPREADER LAZY-L INDV BAG CS500 Catalog #89209-364
Product 89209-364 is similar to Competitor's number 47732-760, but manufacturer and specifications may vary.
VWR CELL SPREADER PP STR PK100 Catalog #89042-020
Product 89042-020 is similar to Competitor's number 47732-760, but manufacturer and specifications may vary.
VWR CELL SPREADER PP STR PK100 Catalog #89042-021
See more alternativesProduct 89042-021 is similar to Competitor's number 47732-760, but manufacturer and specifications may vary.
Specifications
- Cat. no.:47732-760
- Packaging:Individually Wrapped (1 per Pouch)
Specifications
About this item
These durable polystyrene L-shaped spreaders are an alternative to bending glass rods or pipets.
Disposable and nonbreakable, these spreaders do not present a safety hazard. The L-shaped design allows for a smooth and even sample distribution, reducing the likelihood of the spreader digging into the agar. Presterilized to eliminate flaming or autoclaving and reduce the risk of contamination.
Ordering information: Available individually wrapped or 10 per pouch.
Customers Who Bought This Also Bought