360mL Stainless Steel Temp-Control Container for Waring® Blenders, Eberbach
Supplier: Eberbach
E8590.00
58983-048EA 2408.19 CAD
58983-048
360mL Stainless Steel Temp-Control Container for Waring® Blenders, Eberbach
Homogenizers Standard Laboratory Blenders
Stainless steel jacket maintains low or high temperature for improved cutting results or preservation of samples sensitive to temperature rise due to cutting friction or laboratory atmosphere
Fits Waring power units. Cooling/heating is regulated by circulating liquid through tubulatures or inserting ice/dry ice via top opening. Total capacity: 360mL; working volume: 25 to 250mL. Closures provided for all openings.
Ordering information: Supplied with cover, rubber stoppers and blending assembly. Without motor base.