Specifications
- Description:Wedge Pan
- Cat. no.:66104-100
Specifications
About this item
Great for chemical spill clean-up
- Inexpensive
- Disposable
- Convenient to use
Thin-edged wedge pan scoops up the refuse for insertion into the proper disposal receptacle. A single pan is useful in most instances. For large spills, two pans pushed toward one another will gather the spill more quickly. Pan measures 7x71/4" and tapers from a thin edge to 11/4" deep. The recessed hand grip provides room for fingers and thumb.
Use to scoop up spilled chemicals after containment by an absorbent medium.