Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

Methyl red

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
 
 
SearchResultCount:"12"
  List View Easy View
Sort by:
 
 
 
 


Supplier:  AVANTOR PERFORMANCE MATERIALS
Description:   Alcohol Solution
SDS
Catalog Number: (CA11021-642)

Supplier:  VWR
Description:   C.I Acid red 2
SDS
Catalog Number: (CA1.06076.0100)

Supplier:  MilliporeSigma
Description:   Grade Acs,Reagent Ph Eur
Supplier:  TCI America
Description:   CAS Number: 493-52-7
MDL Number: MFCD00002425
Molecular Formula: C15H15N3O2
Molecular Weight: 269.30
Form: Crystal
Color:...
SDS
Catalog Number: (TCM0702-025G)

Supplier:  TCI America
Description:   CAS Number: 493-52-7
MDL Number: MFCD00002425
Molecular Formula: C15H15N3O2
Molecular Weight: 269.30
Purity/Analysis Method: &...
SDS
Supplier:  TCI America
Description:   [for Titration]
CAS Number: 493-52-7
MDL Number: MFCD00002425
Molecular Weight: 269.30
Form: Clear Liquid
Color: Red
SDS
Catalog Number: (TCM0056-500ML)

Supplier:  TCI America
Description:   [for pH Determination]
CAS Number: 493-52-7
MDL Number: MFCD00002425
Molecular Weight: 269.30
Form: Clear Liquid
Color: Re...
SDS

Supplier:  Ricca Chemical
Description:   METHYL RED 1GPL IPA 32OZ
SDS
Small Business Enterprise
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 12
  1  2  Next