Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Methyl red sodium salt

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
 
 
SearchResultCount:"7"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Avantor Performance Materials
Description:   Lot analysis on label.
SDS
Supplier:  MilliporeSigma
Description:   Grade Acs, Cas Number 845-10-3
SDS
Supplier:  Alfa Aesar
Description:   METHYL RED SODIUM SALT 98% 25G
Supplier:  TCI America
Description:   CAS Number: 845-10-3
MDL Number: MFCD00002426
Molecular Formula: C15H15N3O2
Molecular Weight: 291.29
Form: Crystal
Color:...
SDS
Supplier:  Alfa Aesar
Description:   METHL RD SOD SLT .02% . 500ML
Supplier:  Ricca Chemical
Description:   Indicator.
SDS
Small Business Enterprise
Supplier:  Alfa Aesar
Description:   METHYL RED SODIUM SLT ACS 100G
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 7  of 7
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping