Precoated Plastic-Backed TLC Sheets, MilliporeSigma
Supplier: MilliporeSigma
Certificates
About this item
For analytical TLC
F is a fluorescent indicator. F254 is a fluorescent indicator with a 254nm excitation wavelength.
The sorbents used on these plates are carefully standardized and controlled. Each is characterized by narrow particle size and pore volume distribution, standardized surface area and activity, and high resolution and capacity. Precoated products are stable and economical. Feature uniform, hard, abrasion-resistant coating.