Order Entry
ContactUsLinkComponent
VWR® Conical-Bottom Culture Tubes, Plastic, without Caps
VWR® Conical-Bottom Culture Tubes, Plastic, without Caps
Catalog #: 60818-099
Supplier:  VWR International
Sustainable Packaging
VWR® Conical-Bottom Culture Tubes, Plastic, without Caps
Catalog #: 60818-099
Supplier:  VWR International
Sustainable Packaging
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
CENTRIFUGE TUBE CONE 5ML PK250 Catalog #80076-634

Product 80076-634 is the Avantor number for Competitor's number 60818-099.

Specifications

  • Dimensions:
    12×75 mm
  • Volume:
    4 mL
  • Description:
    Polypropylene Tubes
  • Environmentally Preferable:
    Sustainable Packaging,
    Low Manufacturing Impact

Specifications

About this item

Conical-bottom culture tubes are non sterile and fabricated from durable polystyrene or polypropylene.

Delivery: These disposable tubes are not supplied with caps.
Customers Who Bought This Also Bought