Specifications
- Description:E. coli BL21
- Cat. no.:CA95017-592L
Specifications
About this item
The E. coli BL21 strain is protease-deficient for optimal expression of a variety of recombinant proteins.
- High level expression of a variety of recombinant proteins
- Defective in E. coli strain B and wild-type restriction systems to permit high efficiency transformation with unmodified plasmid DNA
- Verified by RAPD fingerprint analysis using ready-to-go RAPD analysis kit
- Supplied lyophilised
Well suited for use with the GST gene fusion system, E. coli BL21 is an exemplary host for the expression of recombinant proteins. As an E. coli B strain, it further lacks the major protease, encoded by the lon gene, catalysing the endoproteolytic cleavage of damaged and recombinant proteins in the cell.
The protease-minus nature of BL21 makes it useful as an expression host. Since BL21 does not transform well, use an alternate strain for cloning and maintenance of the vector.
Genotype: The BL21 strain is derived from E. coli B. The genotype is F-, ompT, hsdS (rB -, mB -), gal, dcm.
Growth conditions: Resuspend lyophilised cultures in 1 ml of LB medium. Grow overnight at 37 °C before plating onto LB agar plates.
Storage conditions: −20 °C. Mix equal volumes of stationary phase culture (grown in LB medium) and glycerol. Store at –70 °C. Revive frozen glycerol stocks of BL21 by streaking onto LB agar plates.