Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "torches"

https://ca.vwr.com/stibo/smallweb/std.lang.all/99/27/18989927.jpg

Portable sources of illumination for maintenance, inspection, or emergency scenarios where traditional power sources have been cut off, flashlights provide a reliable source of independent lighting that is dependable with regular battery changes or recharging. Heavy-duty fabrication materials make these items chemically resistant and capable of absorbing shock from being dropped. Uniform beam of illumination provides a clear line of sight to illuminated objects. Easy to use; depress button to activate light.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"7"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalog Number: (29960-016)
Supplier:  VWR International
Description:   Each flashlight features LED lights, 100,000-hour bulbs, and light that can be seen for up to 2km at full charging capacity.
Limited Availability
Supplier:  BDS ROCKS, LLC.
Description:   Powerful Enough to Fluoresce Samples Even in Daylight
Catalog Number: (470012-760)
Supplier:  Skywill Far East Industries
Description:   Made of Unbreakable Molded Plastic.
Supplier:  ESM INC ENTERPRISES SE
Description:   Science Teachers Make A World of Difference.
Catalog Number: (470145-480)
Supplier:  RAYOVAC CORPORATION
Description:   Heavy Duty Flashlight.
Catalog Number: (470153-336)
Supplier:  MEICO LAMP
Description:   Squeeze Light, LED, Blue
Catalog Number: (470093-426)
Supplier:  MEICO LAMP
Description:   SQUEEZE LIGHT LED RED
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 7  of 7