VACUUBRAND® XS-Series Rotary Vane Vacuum Pumps, BrandTech
Supplier: BrandTech
VACUUBRAND®
698143
80089-634EA 11318.03 CAD
80089-634
VACUUBRAND® XS-Series Rotary Vane Vacuum Pumps, BrandTech
Pumps Vacuum Pumps
These two-stage vacuum pumps are designed especially for chemistry applications
  • Designed from the ground up for laboratory applications
  • Energy-efficient
  • Compact design
  • Quiet and low vibration
The inlet connection for pumps 80089-598 and -630 is a KF16 small flange with an included 10mm (3/8") hose barb; the outlet connection is a 10mm (3/8") hose barb. The inlet and outlet connections for pump 80089-634 use a KF25 small flange.
Note: Always use a cold trap with applications involving solvent vapors.
Many parts are plasma-treated for maximum corrosion resistance. Anti-suckback valve reduces corrosive vapors entering pump. A redesigned oil circuit reduces wear on moving components, and a number of other wearing parts and forces have been reduced for longer life. These compact and energy-efficient pumps are one-third to one-half of the size of other rotary vane pumps of similar capacity, conserving valuable lab space.
Ordering information: Pumps are supplied with plug, operating manual, and one-year manufacturer's warranty. Other voltages are available; contact your VWR representative for more information.