Acrodisc® Serum Syringe Filter, 37 mm, Sterile, Cytiva (Formerly Pall Lab)

Supplier: Cytiva
Acrodisc®
4525 4525
CA28143-295PK 282.44 CAD
28143-295 CA28143-295
Acrodisc® Serum Syringe Filter, 37 mm, Sterile, Cytiva (Formerly Pall Lab)
Filters Syringe Filters
For filtration of sera and other viscous, particulate-laden biologicals.

  • For viscous particulate
  • Two layers
  • Hydrophilic
  • For 20 to 100 ml samples

Incorporates a layer of glass over a layer of Supor® (hydrophilic polyethersulfone) membrane.

Max. operating pressure: 75 psi.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR