Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Accessories

Supplier: VWR International
980360



 

ORDER

Expand All / Collapse All
Description For Use With VWR Catalog Number Unit Price Quantity
Large Vessel Carrier with Four 45.7 cm (18") Bars, Stainless Steel, 76.4L x 45.9W x 35.3H cm (30.1 x 18.1 x 13.9") Large Sample Containers, Carboys, Jugs, and Bottles 97003-606 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description For Use With VWR Catalog Number Unit Price Quantity
50 mL Centrifuge Tube Rack For Seventeen 50 mL Conical Tubes 12985-052 Each Retrieving
Adjustable Platform with Four Bars, 45.7 x 61 cm (18 x 24") - 97003-604 Each Retrieving
Adjustment Bar, Replacement, 45.7 cm (18") - 97003-612 Each Retrieving
Culture Platform with Mat, 45.7 x 61 cm (18 x 24") - 97003-600 Each Retrieving
Double Tier Braces, Set of 4 - 97003-646 Each Retrieving
Microplate Clamp 97003-634 Each Retrieving
Pivoting Test Tube Rack For 16 mm Tubes 97003-638 Each Retrieving
Pivoting Test Tube Rack For 20 mm Tubes 97003-640 Each Retrieving
Pivoting Test Tube Rack For 25 mm Tubes 97003-642 Each Retrieving
Pivoting Test Tube Rack For 30 mm Tubes 97003-644 Each Retrieving
Pivoting Test Tube Rack For 13 mm Tubes 97003-636 Each Retrieving
PVC Clamp For 2 L Erlenmeyer Flasks 97003-584 Each Retrieving
PVC Clamp For 1 L Erlenmeyer Flasks 97003-582 Each Retrieving
PVC Clamp For 500 mL Erlenmeyer Flasks 97003-580 Each Retrieving
PVC Clamp For 250 mL Erlenmeyer Flasks 97003-578 Each Retrieving
PVC Clamp For 125 mL Erlenmeyer Flasks 97003-576 Each Retrieving
Rubber Mat, 45.7 x 61 cm (18 x 24") - 97019-052 Each Retrieving
Stainless Steel Clamp For 125 mL Erlenmeyer Flasks 57018-800 Each Retrieving
Stainless Steel Clamp For 50 mL Erlenmeyer Flasks 57018-797 Each Retrieving
Stainless Steel Clamp For 25 mL Erlenmeyer Flasks 57018-786 Each Retrieving
Stainless Steel Clamp For 10 mL Erlenmeyer Flasks 57018-775 Each Retrieving
Stainless Steel Clamp For 1 L Media Bottles 14215-238 Each Retrieving
Stainless Steel Clamp For 500 mL Media Bottles 14215-236 Each Retrieving
Stainless Steel Clamp For 6 L Erlenmeyer Flasks 14215-232 Each Retrieving
Stainless Steel Clamp For 4 L Erlenmeyer Flasks 14215-230 Each Retrieving
Stainless Steel Clamp For 2.8 L Erlenmeyer Flasks 14215-228 Each Retrieving
Stainless Steel Clamp For 2 L Erlenmeyer Flasks 14215-226 Each Retrieving
Stainless Steel Clamp For 1 L Erlenmeyer Flasks 14215-224 Each Retrieving
Stainless Steel Clamp For 500 mL Erlenmeyer Flasks 57018-822 Each Retrieving
Stainless Steel Clamp For 250 mL Erlenmeyer Flasks 57018-811 Each Retrieving
Test Tube Rack For Forty-Eight 10–14 mm Tubes 14215-240 Each Retrieving
Test Tube Rack For Twenty-One 21–25 mm Tubes 14215-244 Each Retrieving
Test Tube Rack For Thirty-Three 16–20 mm Tubes 14215-242 Each Retrieving
Universal Platform, 45.7 x 61 cm (18 x 24") - 97003-590 Each Retrieving
Universal Platform, 45.7 x 76.2 cm (18 x 30"); Two-Tier Ready - 97003-594 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping