Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Gas Bags
https://ca.vwr.com/stibo/smallweb/std.lang.all/52/72/18945272.jpg
Designed for sampling high-level VOCs as well as light and permanent gases, the gas bags feature valves with a hose connection for attaching tubing. Low-bleed assembly inside the valves prevents sample contamination from the septum. With chemical inertness and resistance to abrasion, the correct film type will meet procedure needs. The gas bags meet a variety of sampling applications including stationary sources emissions, workplace atmospheres, indoor air quality, and breath analysis.
SearchPresentationType-VERTICAL
SearchResultCount:"6"
  List View Easy View
Sort by:
Supplier:  Restek
Description:   Sampling bags are a low-cost whole-air sampling device for high-level VOCs and permanent gases.
Supplier:  Saint Gobain Performance Plastics
Description:   An economical alternative for analysis of noncorrosive gases such as carbon monoxide, sulfur dioxide, hydrogen sulfide, radon, and mercaptans.
Supplier:  Restek
Description:   Replacement septa for sampling bags are available in PTFE-faced silicone formats.
Supplier:  Restek
Description:   ALTEF Gas Sampling Bags are an excellent low-cost alternative to Tedlar bags.
Supplier:  Saint Gobain Performance Plastics
Description:   For gas sample analysis, sampling smokestack exhaust systems, monitoring air quality, and calibrating instruments in the field.
Catalog Number: (10803-304)
Supplier:  Restek
Description:   Sampling bags are a low-cost whole air sampling device for high level VOCs and permanent gases.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 6  of 6