Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Sterilisation Pouches

https://ca.vwr.com/stibo/smallweb/std.lang.all/98/54/18989854.jpg

The sterilization pouches are designed for quick, easy presentation of sterile items. Flat seals ensure seal integrity, and that the bags will not open up in steam autoclaves or be affected by gas. Sterilization pouches facilitate effective disinfection, safe handling, and storage of all items until the moment they are used. The pouches maintain the sterility nature of contents stored inside until the self-sealing, adhesive strips, or heat sealed closure is opened.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"18"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   Sterilization pouches are made of surgical-grade paper and blue polypropylene/polyester film.
Product available on GSA Advantage®
Supplier:  HARDY DIAGNOSTICS CA
Description:   Autoclave Bag, red, with heat indicator, polypropylene 2mil thick, 200 per case, by Medical Action Industries
Supplier:  GOSSELIN NORTH AMERICA/CORNING CA
Description:   Featuring a printed biohazard logo, bags are specifically designed for laboratory waste and decontamination in autoclave applications.
Catalog Number: (89200-840)
Supplier:  GOSSELIN NORTH AMERICA/CORNING CA
Description:   Gosselin™ Autoclave Bags are available in HDPE or Polypropylene formats, with a capacity of 14L
Supplier:  HARDY DIAGNOSTICS CA
Description:   Autoclave Bag, 8x12 inches, red, with Indicator, polypropylene 2mil thick, 400 per case, by Medical Action Industries
Supplier:  Cardinal Health
Description:   Peel-open pouches are designed for quick, easy presentation of sterile items
Supplier:  Cardinal Health
Description:   Ideal for odd-sized, long, or bulky objects
Limited Availability
Supplier:  Propper
Description:   Instant, self-sealing sterilization pouches eliminate the need for heat sealing and separate sealing tapes
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 18
  1  2  3  Next