Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Boiling Stones

https://ca.vwr.com/stibo/smallweb/std.lang.all/49/62/18944962.jpg

Boiling stones are unevenly-shaped substances that are added to liquids to stabilize. Chemically-inert, anti-bumping granules come in a variety of particle sizes. With a porous nature, boiling chips can successfully trap air in cavities to form solvent vapor bubbles continuously as long as the liquid remains at boiling point. Without the stones, liquids have a tendency to superheat then violently boil over. Boiling stones prevent intense reagent splattering, equipment damage, or the experiment being negatively impacted.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"6"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Catalog Number: (CA11021-424)
Supplier:  VWR
Description:   Boiling stone granules, sold in poly bottles.
Catalog Number: (CA1.07912.0250)
Supplier:  MilliporeSigma
Description:   Boiling chips for Kjeldahl digestion.
Supplier:  Saint Gobain Performance Plastics
Description:   Stones promote gentle, efficient boiling by creating many projections on which small bubbles can form
Catalog Number: (26397-409)
Supplier:  Walter Stern
Description:   Treated, micro-porous, chemically inert carbon chips eliminate “bumping” in either aqueous or non-aqueous boiling liquids
SDS
Supplier:  Hengar
Description:   High purity, white, amphoteric alundum granules
Small Business Enterprise
Supplier:  Walter Stern
Description:   VWR GLS BEADS SOLID 6MM 1LB/PK.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 6  of 6