Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • GSA Contract (9)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "centrifuge-bottles"

https://ca.vwr.com/stibo/smallweb/std.lang.all/50/14/18945014.jpg

Centrifuge bottles are manufactured from polycarbonate or polypropylene, giving the containers translucent qualities for easy identification of solutions within. With wide mouths, the containers fill and decant materials easier as well as permit easy retrieval. Sealing caps and bottles are autoclavable. Centrifuge bottles with gradations or unique shapes will assist in sediment concentration at the bottom of the centrifuge bottle or aid in cell culture applications.

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"18"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   Centrifuge bottles are manufactured from polycarbonate or polypropylene.
Supplier:  Thermo Scientific
Description:   Wide mouth bottles are translucent and have excellent chemical resistance. All caps are polypropylene. Sealing caps feature a retained silicone gasket...
Supplier:  Beckman Coulter
Description:   Bottle assemblies are for use in high-performance and high-capacity rotors.
Supplier:  Beckman Coulter
Description:   Beckman Coulter one-liter bottles build upon the versatility of the Avanti Series of centrifuges to help users gain even more control of workflow.
Supplier:  DWK Life Sciences (KIMBLE®)
Description:   Bottle features a small bottom area for better concentration of sediment, which facilitates decanting of liquid. Supplied with a white polypropylene l...
Supplier:  Thermo Scientific
Description:   Wide mouth bottles are transparent, with excellent mechanical strength. All caps are polypropylene except the cap for bottle 89050-118, which i...
Supplier:  CORNING CA
Description:   For use with relatively small amounts of solids. Bottles feature a small bottom that permits sediment to be concentrated for decanting operations. Rub...
Product available on GSA Advantage®
Supplier:  Thermo Scientific
Description:   Ideal for use in refrigerated centrifuges. Wide mouth provides easy access. Withstands temperatures to –100°C (–148°F). Manufactured from chemically r...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 18
  1  2  3  Next