Would you like to log in again?
  • Criteria
    • GSA Contract (54) SBE (27)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Burettes
https://ca.vwr.com/stibo/smallweb/std.lang.all/68/44/20236844.jpg
Burets are devices used typically in analytical, quantitative chemistry applications for measuring liquid solution or gases. Differing from a pipette since the sample quantity delivered is changeable, graduated burets are used heavily in titration experiments. In liquid measurement styles, the stopcock preventing leaks is located at the bottom of the vertical glass tube, while for gas models it is located at the top. For the most precise readings, digital options are available.
SearchPresentationType-VERTICAL
SearchResultCount:"90"
  List View Easy View
Sort by:
Supplier:  VWR International
Description:   These hardy burets are ideal for classroom use.
Supplier:  VWR International
Description:   Borosilicate glass buret for general industrial applications.
Supplier:  VWR International
Description:   Borosilicate glass buret assembly with precision-grade white scale.
Supplier:  VWR International
Description:   Provides accurate and reliable titration results with the convenience of a large digital display, eliminating tedious meniscus readings.
Supplier:  VWR International
Description:   Borosilicate glass burets for use with light-sensitive liquids.
Supplier:  VWR International
Description:   Precision-grade borosilicate glass buret assembly and receiving bottle.
Supplier:  VWR International
Description:   Made from borosilicate 3.3 expansion glass.
Catalog Number: (17477-020)
Supplier:  VWR International
Description:   Borosilicate glass buret manufactured to Class A specifications for burets.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 90
  1  2  3  4  5  6  7  8  9  10  11  12  Next 