Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • Environmentally Preferable (4) GSA Contract (5) SBE (7) UL Listed (12)
  • Supplier
  • Refine by Suppliers
    Sort by:

Safety Cans

https://ca.vwr.com/stibo/smallweb/std.lang.all/97/56/18989756.jpg

Holding corrosives or flammable liquids such as paint thinner and lighter fluid, the leak-proof safety cans are designed to comply with safety codes and regulations. Easy-to-fill designs with tight seal closures provide maximum protection for the environment and employees. Able to withstand physical shock encountered during transportation, safety cans store and dispose solvents safely. Select models feature a hose for direct pouring or carbon filters that trap chemical vapors from being inhaled.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"39"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   This solvent waste system is designed to be an affordable solution for HPLC liquid waste disposal.
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
Catalog Number: (13197-284)

Supplier:  Bel-Art Products
Description:   For safe, in-plant handling and temporary storage of spent solvents or other liquid wastes awaiting proper disposal.
Catalog Number: (10061-000)

Supplier:  S.C.A.T
Description:   HDPE
Catalog Number: (10811-224)

Supplier:  New Pig
Description:   Recycling system easily punctures aerosol cans, rendering them exempt from hazardous waste regulations when properly emptied.
Supplier:  Justrite
Description:   Safety Can for Liquid Disposal, Material: HDPE, with Stainless steel hardware, built-in fill guage no. 08530, Material: Polyethylene, Colour: Red, Dimension Exterior: 14.75in H x 12in OD, Capacity: 2 gal (7.5L), Fill/Pour Hole: 1.45in, Vent: 3 and 5 psig
Supplier:  Qorpak
Description:   Epoxy Lined Round Paint Can with Triple Tite Lid provide a non-reactive barrier between the container and the product/atmosphere; protecting the conte...
Product available on GSA Advantage® Small Business Enterprise
Supplier:  Justrite
Description:   High-density polyethylene cans for containment of corrosives and liquid flammable wastes.
Supplier:  Justrite
Description:   Compliant and economical Justrite™ Type I safety cans reduce fire risks and make everyday use of flammables easier and safer.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 39
  1  2  3  4  5  Next