Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • GSA Contract (17) SBE (2)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

Bioreactors

https://ca.vwr.com/stibo/smallweb/std.lang.all/68/38/20236838.jpg

Bioreactors are apparatuses that support active biological reactions or processes in a research environment. Granting optimal metabolic activity and cell or tissue growth for harvesting, bioreactors also support close chemical aerobic or anaerobic process examinations. By superior use of valuable space, the tool reduces repetitious manual handling. Chamber openings allow for an even fill for consistent results within the vessel. Specialized membrane technology causes the best separation between the cell cultivation area and the media chamber.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"13"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   This sterile cell culture chamber, which is available with 1, 2, 5, or 10 layers, produces large-scale cell culture results in just one cycle.
Supplier:  Corning
Description:   Corning CELLine Disposable Bioreactor for antibody and recombinant protein production.
Supplier:  Thermo Scientific
Description:   Highly versatile and easy to use, the EasyFill Cell Factory system offers a sterile, single-use solution designed to optimize valuable incubator space...
Supplier:  Thermo Scientific
Description:   Cell factory units maximize available growing area with low risk of contamination and easy handling
Supplier:  Thermo Scientific
Description:   Closure with overhead mixer support for use with Closed-Dome Bio Tanks (see 66463-206 series)
Supplier:  Thermo Scientific
Description:   Excellent for preparing media components and dispensing and growing cultures
Supplier:  CORNING CA
Description:   The CellSTACK® culture chamber features a 1,272cm² cell growth area and two 26mm diameter filling ports allowing direct access to chamber bottom
Product available on GSA Advantage®
Catalog Number: (89090-252)

Supplier:  CORNING CA
Description:   The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment
Product available on GSA Advantage®
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 13
  1  2  Next 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping