Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Product Category
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "sleeve-protectors"

https://ca.vwr.com/stibo/smallweb/std.lang.all/97/99/18989799.jpg

Manufactured from microporous fabrics to be exceptionally clean and durable, the sleeve protectors serve as fluid and particulate barriers. These breathable garments provide resistance to common chemicals encountered, making the apparel versatile for use in any testing application. Sleeve covers feature serged seams and elastic wrists and tops for easy fitting and universal comfort. Models built with an emphasis in chemical protection, offer excellent resistance against biohazards such as blood, body fluids, and viral contaminants.

SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-SearchPresentationType-VERTICAL
 
 
SearchResultCount:"43"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   VWR® Advanced Protection Sleeves are manufactured from a specially formulated breathable microporous fabric that is exceptionally clean and durable.
Supplier:  VWR International
Description:   VWR® Sterile Sleeves are manufactured from a specially formulated breathable microporous fabric that provides significant fluid and barrier protection...
Catalog Number: (89208-758)
Supplier:  VWR International
Description:   Disposable non-woven sleeves are made of durable 30g SPP fabric.
Supplier:  VWR International
Description:   Sleeve covers are constructed of DuPont™ Tyvek® nonwoven spunbonded olefin for premium protection in cleanroom environments while remaining comfortabl...
Limited Availability
Supplier:  VWR International
Description:   Sleeves provide reliable protection in laboratory, electrical, pharmaceutical, food, industrial, and cleanroom applications.
Catalog Number: (CA10010-136)
Supplier:  DuPont
Description:   Tyvek® garments are composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
Supplier:  Kimberly-Clark
Description:   Ideal for food processing and assembly operations. Low-linting MICROFORCE™ Barrier SMS fabric sleeve protectors provide protection against dry particu...
Catalog Number: (CA13452-680)
Supplier:  DuPont
Description:   DuPont™ Tychem® QC is a comfortable, lightweight, and durable fabric that is designed for light splash protection in a variety of industrial environme...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 43
  1  2  3  4  5  6  Next