Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • SBE (1)
  • Supplier
  • Refine by Suppliers
    Sort by:

Instrument Trays/Dishes

https://ca.vwr.com/stibo/smallweb/std.lang.all/94/42/18989442.jpg

Rectangular instrument trays and dishes are available in shallow platform or deep well styles for desired contents or use. Organizing required surgical tools, the smooth-surfaced holders allow personnel to quickly access the correct device when time matters most. Perforated options permit steam penetration to tools during autoclaving and allow water to escape for basic air drying. The instrument trays and dishes safely hold and transport multiple pieces to cabinets or refrigerated storage.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"59"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  VWR International
Description:   Recyclable trays feature a built-in angle to prevent separation of thermometer liquid.
Supplier:  Bel-Art Products
Description:   Trays are steam autoclavable at 121°C (250°F).
Supplier:  Thermo Scientific
Description:   Conforms to NIH and Federal Register guidelines for biohazardous and recombinant DNA research, where disinfecting and subsequent autoclaving of pipets...
Catalog Number: (10193-250)

Supplier:  Sklar
Description:   Solid Pans are constructed of stainless steel
Catalog Number: (75817-522)

Supplier:  Sklar
Description:   Minor Surgery Trays are industry standard
Supplier:  Sklar
Description:   Solid, stainless steel instrument trays
Supplier:  Sklar
Description:   Instrument Tray, Solid, Heavyweight
Supplier:  Sklar
Description:   Plastic Procedure Tray
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 59
  1  2  3  4  5  6  7  8  Next