Would you like to log in again?
  • Criteria
    • GSA Contract (95)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Quantitative Filter Paper
SearchPresentationType-VERTICAL
SearchResultCount:"30"
  List View Grid View Easy View (new)
Sort by:
Supplier:  VWR International
Description:   Use for retention of zirconium hydroxide, aluminum hydroxide, cobalt sulfide, and other coarse or gelatinous precipitates.
Supplier:  VWR International
Description:   Useful for very fine precipitates such as barium and lead sulfates, stannic and nickel sulfides, and calcium hydroxide.
Product available on GSA Advantage®
Supplier:  GE Healthcare - Whatman
Description:   For coarse particles
Supplier:  GE Healthcare - Whatman
Description:   GE’s Whatman Grade 42 quantitative papers have the finest retention of all Whatman ashless filter paper grades
Supplier:  GE Healthcare - Whatman
Description:   Whatman Grade 40 high-purity filters from GE Healthcare’s Life Sciences business are general-purpose papers for quantitative analysis. Grade 40 filter...
Supplier:  GE Healthcare - Whatman
Description:   GE’s Whatman Grade 41 environmental pollution papers are among the fastest filter papers in the Whatman ashless range. This gravimetric filter is well...
Supplier:  GE Healthcare - Whatman
Description:   GE’s Whatman Grade 541 acid hardened cellulose filter paper features high wet-strength, high chemical resistance, and a large nominal particle retenti...
Supplier:  GE Healthcare - Whatman
Description:   Grade 802 prepleated filter paper for use with a conical filter funnel, offers fast filtration and high loading capacity for analysis involving coarse...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 30
  1  2  3  4  Next