Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • GSA Contract (35)
  • Supplier
  • Refine by Suppliers
    Sort by:

Membrane Filter Capsules

https://ca.vwr.com/stibo/smallweb/std.lang.all/95/13/18989513.jpg

Designed for fast processing and final filtration of aqueous samples, capsule filters provide efficient processing and enhanced throughput of difficult-to-filter solutions via an in-line installation. Applications include large-volume media preparation, pilot-scale manufacturing, and sterile filtration of buffers or biological solutions. These capsules are suitable for point-of-use filtration for laboratory water. Capsules are sterilized by gamma irradiation prior to bagging and the polycarbonate/polypropylene housing in most models ensure easy, rapid installation with no maintenance required.


SearchPresentationType-VERTICAL
 
 
SearchResultCount:"26"
  List View Easy View
Sort by:
 
 
 
 

Supplier:  Pall Laboratory
Description:   Designed for fast processing and final filtration of aqueous samples.
Supplier:  Pall Laboratory
Description:   Economical, disposable capsule filters feature either a Supor® (hydrophilic polyethersulfone) membrane for high throughput processing or a Fluorodyne®...
Supplier:  Pall Laboratory
Description:   Polypropylene prefilter is designed for biologicals, serum-based products, beverages, plating solutions, ink, syrup, water, and a wide range of solven...
Catalog Number: (CA28145-145)

Supplier:  Pall Laboratory
Description:   Designed to provide bacteria-free air when required for sterile applications
Supplier:  Pall Laboratory
Description:   Designed for collection and recovery of waterborne contaminants, Cryptosporidium, and Giardia, with typically greater than 70% recovery of target orga...

Supplier:  Pall Laboratory
Description:   These 100% integrity-tested filters feature high flow rate venting and aggressive solvent filtration.
Supplier:  Pall Laboratory
Description:   Supracap 50 depth filter capsules are ideal prefilters made with Seitz® high performance depth filter media for the removal of unwanted solids from so...
Supplier:  Pall Laboratory
Description:   Membrane capsule is designed to filter liquids not requiring sterilization, or as a prefilter before final sterilization.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address


VWR will not lift restrictions for residential shipping addresses.

-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 26
  1  2  3  4  Next