Would you like to log in again?
  • Criteria
    • SBE (29) CSA Certified (7)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
You Searched for: "eyewash-stations-and-eyewash-bottles"
https://ca.vwr.com/stibo/smallweb/std.lang.all/52/13/18945213.jpg
Eyewash stations for quick and accessible care in emergencies are offered at VWR. Open, wall mounted models feature bottles of sterile saline with twist-off tops and flexible eye cups; select closed stations for dirt and dust prone work spaces. Delivery systems offer an automated, gravity-fed and hands-free solution without plumbing. Convert your lab faucet to an eyewash station with a faucet mount unit that attaches with a threaded inlet. Plumbed eyewash stations that require a dedicated water source are also available.
SearchPresentationType-VERTICAL
SearchResultCount:"46"
  List View Grid View Easy View (new)
Sort by:
Supplier:  Bel-Art
Description:   This easily recognized, ready-to-use Eye Wash Station comes complete with eye wash bottle, dust cover, mirror, and instructions for use and service.
Catalog Number: (VF-0927-SE)
Supplier:  Speakman
Description:   The eyewash features a stay-open valve and twin aerated sprays with flip-top dust caps and automatic flow control. Designed for countertop mounting, t...
Catalog Number: (45500-180)
Supplier:  Encon Safety Products
Description:   HYDROSEP ADDITIVE 8OZ CS4.
Supplier:  WaterSaver Faucet
Description:   Constructed of chrome-plated brass, these faucet-mount units convert any laboratory faucet into an emergency eyewash station without special plumbing ...
Supplier:  Speakman
Description:   For use in areas where workers could be exposed to harmful contaminants. Eyewashes flush liquid and dust contaminants from the eyes when activated by ...
Supplier:  Speakman
Description:   The SE-4400 portable eyewash allows to locate an eyewash anywhere that it is needed, even where potable water is not available.
Catalog Number: (CA82004-272)
Supplier:  Brady Worldwide
Description:   Our Double Bottle Eye Wash Stations feature a drop down front panel that allows quick access to your eyewash bottles (not included). Proper usage inst...
Supplier:  Honeywell Safety Products
Description:   This highly visible green station delivers purified, contaminant-free saline solution from a sealed, integrated delivery module.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
You must log in to order restricted items. We request that you provide the required business documentation to purchase this product for the first time.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • issued document with your organization's Federal Tax ID Number
  • Government issued document with your organization's Resale Tax ID Number
  • Any other Government ID that includes the business name and address
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is currently unavailable but limited stock may be available in our extended warehouse network. Please call 1-800-932-5000 and a VWR Customer Service Representative will help you.
1 - 8  of 46
  1  2  3  4  5  6  Next